S-Nitrosoglutathione (GSNO) ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-(nitrosothio)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, GSNO, Nitrosoglutathione, S-Nitroso-L-Glutathione

420-002-M025 420-002-M100
89150-690EA 197.5 CAD
89150-690 89150-692
S-Nitrosoglutathione (GSNO) ≥98% (by TLC)
S-Nitrosoglutathione (GSNO)

Nitric oxide donor


NO donor which spontaneously releases NO under physiological conditions. Half-life of 159 hours under physiological conditions. Selectively inhibits platelet activation and displays cardioprotective effects. Inhibits NF-kappaB activation. Inhibits endothelial cell proliferation and induces apoptosis in T cells.

Formula: C₁₀H₁₆N₄O₇S
MW: 336.319 g/mol
MDL Number: MFCD00214341
CAS Number: 57564-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9c8af377d46fbd7b083bb7a00d8fa34e3fc541e&vid=b272f17adfb9c91e4cc583e909347ab318045f50&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR