Teniposide ≥97% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
(5R,5aR,8aR,9S)-9-(((2R,4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-(thiophen-2-yl)hexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
400-053-M025
400-053-M100
89150-676EA
124.82
CAD
89150-676
89150-678
Teniposide ≥97% (by HPLC)
Teniposide
Topoisomerase II inhibitor
Glycoside-derivative of podophyllotoxin. Has cytostatic effect. Inhibits topoisomerase II. Induces apoptosis.
Semisynthetic derivative of podophyllotoxin.
Formula:
C₃₂H₃₂O₁₃S MW: 656.66 g/mol |
MDL Number:
MFCD00866516 CAS Number: 29767-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...