Cyclophosphamide monohydrate (synthetic) ≥97% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
Synonyms:
2-(Bis(2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide hydrate
, Cyclophosphamide hydrate
, 2-[Bis(2-chloroethyl)amino]tetrahydro-1,3,2-oxazaphosphorin-2-oxide
Alkylates DNA, forming DNA-DNA cross-links that result in inhibition of DNA synthesis and cell death
- DNA alkylating agent
- Source: synthetic
- Appearance: White to off-white crystalline solid
- Soluble in water; slightly soluble in 100% ethanol
Interestingly the repair enzyme O6-alkylguanine-DNA alkyltransferase, long thought to be involved with resistance to methylating and chloroethylating agents, may also contribute to resistance to the cytotoxic and mutagenic effects of cyclophosphamide.
Formula:
C₇H₁₇Cl₂N₂O₃P MW: 279.1 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00149395 CAS Number: 6055-19-2 UN: 3464 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...