Mitoxantrone dihydrochloride (synthetic) ≥97%

Supplier: Enzo Life Sciences

Synonyms: Novantrone, 1,4-Dihydroxy-5,8-bis((2-((2-hydroxyethyl)amino)ethyl)amino)anthracene-9,10-dione dihydrochloride

400-050-M010 400-050-M050
89150-664EA 71.1 CAD
89150-664 89150-666
Mitoxantrone dihydrochloride (synthetic) ≥97%
Mitoxantrone dihydrochloride

Antiviral, antibacterial, antiprotozoal, immunomodulating, and antineoplastic cytostatic anthraquinone derivative


  • Blue to black crystalline solid
  • Soluble in water; slightly soluble in 100% ethanol or methanol; insoluble in acetonitrile or acetone


Induces DNA damage by intercalating into DNA and inhibiting topoisomerase II. Induces interstrand DNA cross-links and DNA-protein cross-links in cellular systems. Has been shown to be an inhibitor of DNA methylation.

Formula: C₂₂H₃₀Cl₂N₄O₆
MW: 517.404 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00242943
CAS Number: 70476-82-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e19d12dbe6c82a7d206bd1e571b4086c275bf3fa&vid=b13dbba11552b948c7240f6b8f29dc33eace7b25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR