Oxaliplatin ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

Synonyms: (trans-1,2-Cyclohexanediamine)oxalatoplatinum(II), Oxalato(trans-l-1,2-cyclohexanediamine)platinum(II)

400-042-M005 400-042-M025
89150-642EA 237 CAD
89150-642 89150-644
Oxaliplatin ≥98% (by HPLC)
Oxaliplatin

Antitumor agent.


  • Formulation: White to off-white crystalline powder.


Platinum analog which has wide spectrum anticancer activity. It is found to be more active against human melanoma cell lines, and has better biochemical, pharmacological and cytotoxic properties than cisplatin and carboplatin.


Slightly soluble in water (7.9 mg/mL); very slightly soluble in methanol; almost insoluble in 100% ethanol.

Formula: C₈H₁₄N₂O₄Pt
MW: 397.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866327
CAS Number: 61825-94-3
Merck Index: 14,06912
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609c22c448c18d9a0defc60c94554a21bcd35c61&vid=184482f9de33382d35a45e08fb0328d4ec11f717&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR