Carboplatin ≥98% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
Synonyms:
cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)
400-041-M025
400-041-M100
89150-636EA
83.74
CAD
89150-636
89150-638
Carboplatin ≥98% (by HPLC)
Carboplatin
Antitumor agent.
- Formulation: White to off-white powder.
An analog of cisplatin.
Soluble in water.
Synonym: cis-Diammine [1,1-cyclobutanedicarboxylato] platinum(II)
Formula:
C₆H₁₂N₂O₄Pt MW: 371.26 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00070464 CAS Number: 41575-94-4 EINECS: 255-446-0 Merck Index: 14,01822 |
Specification Test Results
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...