cis-Platin ≥98%

Supplier: Enzo Life Sciences

Synonyms: Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride

400-040-M050 400-040-M250
89150-632EA 94.8 CAD
89150-632 89150-634
cis-Platin ≥98%
cis-Platin

Antitumor agent


  • Formulation: Off-white to orange powder.
  • Endotoxin-content: ≤1.0EU/mg


A potent platinum-based antineoplastic drug. Its mode of action invovles formation of inter- and intrastrand DNA adducts which activate signaling pathways culminating in apoptosis. Platinated DNA adducts also enhance poisoning of DNA topoisomerase I. Cisplatin synergizes with a variety of anticancer agents. Tumor cells rapidly develop resistance to cisplatin and numerous agents are available to block or reverse resistance.

Formula: H₆Cl₂N₂Pt
MDL Number: MFCD00011623
CAS Number: 15663-27-1
UN: 3288
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b290dcdae927c17f1cae98b66a7e5ec5d1ddc50f&vid=8294a97be8c6b8bc0e2527a0a31c865b61b858b3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR