MGD sodium salt monohydrate ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: N-(Dithiocarbamoyl)-N-methyl-D-glucamine.Na.H₂O

400-014-M050 400-014-M250
89150-586EA 124.82 CAD
89150-586 89150-588
MGD sodium salt monohydrate ≥98% (by HPLC)
MGD sodium salt monohydrate

Spin trapping reagent.


  • Formulation: White to off-white solid.


Together with FeSO₄ MGD is a useful component for the formation of the MGD2-Fe²⁺ complex, which is an excellent nitric oxide (NO) spin-trapping reagent. The MGD2-Fe²⁺ complex is quite unstable, especially in the presence of dissolved oxygen. Thus, the complex should be used immediately after being made. An excess (usually 5-fold excess), of MGD to Fe²⁺ is used for making the complex with FeSO₄ to give a more stable complex solution. Acidic conditions should be avoided because dithiocarbamate tends to decompose forming toxic carbon disulfide.


Soluble in water.


Synonym: N-(Dithiocarbamoyl)-N-methyl-D-glucamine.Na.H₂O, N-methyl-D-glucamine dithiocarbamate

Formula: C₈H₁₆NO₅S₂Na·H₂O
MW: 311.3 g/mol
Storage Temperature: Refrigerator
CAS Number: 91840-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9828c4f96012108a955385dfa09a06c4566293b&vid=8261c171e3ba43bab15a5525c1182f152dd43d4c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR