Ciprofloxacin ≥98%

Supplier: Enzo Life Sciences

380-287-G005 380-287-G025
89150-358EA 71.1 CAD
89150-358 89150-360
Ciprofloxacin ≥98%
Ciprofloxacin

Fluoroquinolone antibacterial agent


  • Effective against Mycobacterium tuberculosis
  • Potentially anxiogenic


Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria.


White to off-white powder


Soluble in dilute hydrochloric acid; insoluble in water


Packaging: Shipping temperature: Ambient

Formula: C₁₇H₁₈FN₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00185755
CAS Number: 85721-33-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65d8541ebe2d979e68d209983e742b2a0d2bf269&vid=1b2ede6fcab4038c7211b4cac6a7dd26f434396e&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR