Cyclosporin D
Supplier: Enzo Life Sciences
380-284-M001 380-284-M005
89150-350EA 203.82 CAD
89150-350 89150-352
Cyclosporin D
Proteins and Peptides
TPA/PMA inhibitor
Weak immunosuppressor. Potent inhibitor of tumor promoting phorbol ester TPA/PMA (Prod. No. BML-PE160) in vivo. Potent inhibitor of Ca2+/calmodulin dependent EF-2 phosphorylation in vitro.
Isolated from Fusarium solani.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72b3d1744b46d34a7635af9bfa0d5947da6a8437&vid=a4d46e8b5b34c1fde812e8d214398cb30c0d6098&prid=web&clid=SAPDAM