Vancomycin hydrochloride (from Streptomyces lysosup)
Supplier: Enzo Life Sciences
Synonyms:
(3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
Glycopeptide antibiotic
Interferes with cell wall synthesis. Effective against Gram-positive bacteria.
White to light brown powder
Soluble in water (50 mg/mL)
Packaging: Shipping temperature: Ambient
Formula:
C₆₆H₇₅Cl₂N₉O₂₄·HCl MW: 1485.71 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03613611 CAS Number: 1404-93-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...