Streptomycin sulfate (from Streptomyces species)

Supplier: Enzo Life Sciences

380-277-G100
89150-338EA 157 CAD
89150-338
Streptomycin sulfate (from Streptomyces species)
Streptomycin sulphate

Antibiotic


Inhibits prokaryote protein synthesis. Binds to the S12 protein of the 30S ribosomal subunit. Effective against Gram-negative and Gram-positive bacteria. Mutation in rpsL (the gene for S12 ribosomal protein) prevents binding of streptomycin to ribosome. Inactivated by aminoglycoside phosphotransferase.


White to off-white powder


Soluble in water (50 mg/mL)


Packaging: Shipping temperature: Ambient

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037023
CAS Number: 3810-74-0
https://ca.vwr.com/stibo/web/std.lang.all/05/54/45390554.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR