Doxycycline hyclate ≥98% (dried basis)
Supplier: Enzo Life Sciences
380-273-G001
380-273-G005
89150-324EA
74.26
CAD
89150-324
89150-326
Doxycycline hyclate ≥98% (dried basis)
Doxycycline hyclate
MMP inhibitor
Broad spectrum antibiotic and bacteriostatic. Shows antiprotozoal properties. Potent inhibitor of MMPs (matrix metalloproteinases) in vivo. Inhibits collagen synthesis.
Semisynthetic from oxytetracycline.
Formula:
C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O |
MDL Number:
MFCD07357237 CAS Number: 24390-14-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...