Carbenicillin disodium salt ≥92%

Supplier: Enzo Life Sciences

Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

380-270-G001 380-270-M250
89150-314EA 246.48 CAD
89150-314 89150-316
Carbenicillin disodium salt ≥92%
Carbenicillin disodium salt

Analog to ampicillin


  • Semisynthetic from penicillin


Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-positive and Gram-negative bacteria and Pseudomonas.


White to off-white powder


Soluble in water (50 mg/mL)


Packaging: Shipping temperature: Ambient

Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077683
CAS Number: 4800-94-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e05f545efe4425d010bf97d5c6c1174903a88683&vid=770967cec58fbbae6ca01de4cd1c66f85e52563a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR