Cycloheximide ≥95% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,

380-269-G001 380-269-G005
89150-310EA 123.24 CAD
89150-310 89150-312
Cycloheximide ≥95% (by TLC)
Cycloheximide

Protein synthesis inhibitor


Antibiotic. Inhibits protein synthesis in eukaryotes by interfering with the translocation step. Induces apoptosis in tumor cell lines but can also delay or inhibit apoptosis induced by other agents. Widely used for selection of resistant strains of yeast and fungy, controlled inhibiton of protein synthesis for detecton of short-lived proteins and super-induction of protein expression. At 10µg/ml cycloheximide totally inhibits protein synthesis in mouse thymocytes.


Isolated from Streptomyces griseus.

Formula: C₁₅H₂₃NO₄
MW: 281.35
MDL Number: MFCD00082346
CAS Number: 66-81-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60efd19247841580ab63fad6290efc7a63263e4b&vid=5dd61c17cc66f005734c08abe211814df372286f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR