Thiostrepton

Supplier: Enzo Life Sciences

380-261-G001 380-261-G005
89150-282EA 237 CAD
89150-282 89150-284
Thiostrepton
Thiostrepton

Elongation factor G inhibitor


Thiazole-containing peptide antibiotic. Inhibits protein synthesis by preventing binding of GTP to the 50S ribosomal subunit. Inhibits binding of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome.


Isolated from Streptomyces azureus.

Formula: C₇₂H₈₅N₁₉O₁₈S₅
MDL Number: MFCD00135828
CAS Number: 1393-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b112311ce543cd26cd35950bd76da44b1c03ed2&vid=5f5f3d81bc33b46d889dc2c60d220cf0630dbcec&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR