Antimycin A (from Streptomyces species) ≥90% (by HPLC)

Supplier: Enzo Life Sciences

380-075-M005 380-075-M010
89149-956EA 72.68 CAD
89149-956 89149-958
Antimycin A (from Streptomyces species) ≥90% (by HPLC)
Antimycin A

Antibiotic


  • Mixture of antimycin A's


Induces apoptosis, which is not prevented by the presence of Bcl-2. Inhibits mitochondrial electron transport specifically between cytochromes b and c1.


White to faint yellow powder


Soluble in 100% ethanol (50 mg/mL)


Packaging: Shipping temperature: Ambient

Formula: C₂₈H₄₀N₂O₉
Storage Temperature: Freezer
MDL Number: MFCD01779723
CAS Number: 1397-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f754ba88f3f19ae23ba262dfd93c5ec533ba08d&vid=615cc48e20e325cd83f314364ccf617e3f226e38&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR