Cytochalasin A ≥98% (by HPLC)

Supplier: Enzo Life Sciences

380-057-M001 380-057-M005 380-057-M010
89149-908EA 124.82 CAD
89149-908 89149-910 89149-912
Cytochalasin A ≥98% (by HPLC)
Cytochalasin A

HIV-1 protease inhibitor


Antibiotic. Fungal toxin. Inhibits glucose transport, actin polymerisation and blocks the formation of microtubuli. Inhibits cell division. Inhibits HIV-1 protease.


Isolated from Drechslera dematoidea.

Formula: C₂₉H₃₅NO₅
MDL Number: MFCD00005935
CAS Number: 14110-64-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcfbd95a92322d7670b0ef1e3e2c7add9e520a3f&vid=8eb0d3d0fef805cf81c1ad6f11d3350fc0d30a1f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR