Verruculogen ≥96% (by HPLC, TLC)

Supplier: Enzo Life Sciences

Synonyms: NA 209A

380-056-M001 380-056-M005
89149-904EA 292.3 CAD
89149-904 89149-906
Verruculogen ≥96% (by HPLC, TLC)
Verruculogen

Cell cycle inhibitor


Neurotoxin that acts as an inhibitor of the M phase of the mammalian cell cycle. Inhibits Ca2+-activated K+ channels. Decreases GABA levels in CNS.


Isolated from Penicillium verruculosum.

Formula: C₂₇H₃₃N₃O₇
CAS Number: 12771-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a7e776047f2bfd48b90125898abe06de9c978e6&vid=cf9f23a38a22de60aceaf8802ebbd6adc0a62419&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR