Concanamycin A (from Streptomyces species) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

380-034-C025 380-034-C100 380-034-M001
89149-856EA 527.72 CAD
89149-856 89149-858 89149-860
Concanamycin A (from Streptomyces species) ≥98% (by HPLC)
Concanamycin A

White to off-white solid (sub-milligram quantities will appear as clear film)


Antibiotic. More potent and specific H+-ATPase inhibitor than bafilomycin A1. Inhibits acidification of organelles such as lysosomes and the Golgi apparatus. Blocks cell surface expression of viral glycoproteins without affecting their synthesis. Exhibits cytotoxic effects in a number of cell lines in a cell viability assay. Induces nitric oxide (NO) production. Inhibits Autophagy.


Soluble in methanol, DMSO or acetonitrile, insoluble in water


Packaging: Shipping temperature: Ambient

Formula: C₄₆H₇₅NO₁₄
Storage Temperature: Freezer
CAS Number: 80890-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf747f701fe5a444c8c0c0cb9ad6e554c1644414&vid=5eb87c83ddee690d975e661d3a48987525ef3ad0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR