Monensin monosodium salt ≥90%

Supplier: Enzo Life Sciences

Synonyms: Monensin, Sodium Salt, Monensin sodium salt

380-026-G001 380-026-M100
89149-850EA 151.68 CAD
89149-850 89149-852
Monensin monosodium salt ≥90%
Monensin monosodium salt

Na+ ionophore


Antibiotic that functions as Na+ ionophore. Blocks glycoprotein secretion. Inhibits proliferation of several lymphoma cell lines through cell cycle arrest and apoptosis. Blocks ceramide transport through the Golgi apparatus.

Formula: C₃₆H₆₁NaO₁₁
MW: 692.85 g/mol
MDL Number: MFCD00077826
CAS Number: 22373-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bdc5e1120806cca4d79194ae6d0bd93f4da29eb&vid=a27d584415f73ad5dc2ffc53dfe5bec2ce7135c6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR