Tobramycin (from Streptomyces tenebarius)

Supplier: Enzo Life Sciences

380-018-G001 380-018-M250
89149-834EA 474 CAD
89149-834 89149-840
Tobramycin (from Streptomyces tenebarius)
Tobramycin

White to off-white powder


Aminoglycoside antibiotic with similar structure to the kanamycins and the gentamycins. Used in the treatment of respiratory infection and cystic fibrosis.


Free base


Soluble in water, very slightly soluble in 100% ethanol


Packaging: Shipping temperature: Ambient

Formula: C₁₈H₃₇N₅O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00077885
CAS Number: 32986-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=302747ea1e90f48004af78c991187e52d958f75c&vid=6f889a2c2bd502c008cdafd1a75e5b0c03aa4907&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR