G418 disulfate (from Micromonospora rhodorangea)
Supplier: Enzo Life Sciences
Synonyms:
G418 disulfate, G-418 sulfate, G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate, G-418 disulfate
White to off-white powder
Gentamycin-class aminoglycoside antibiotic. Cytotoxic to prokaryotic and eukaryotic cells. Induces apoptosis. Widely employed in the selection of eukaryotic expression vectors, in combination with either aminoglycoside phosphotransferase 3' or APH II.
Soluble in water
Packaging: Shipping temperature: Ambient
Formula:
C₂₀H₄₄N₄O₁₈S₂ MW: 692.72 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00058314 CAS Number: 108321-42-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...