Tacrolimus ≥98%, crystalline powder

Supplier: Enzo Life Sciences
Danger

380-008-M001 380-008-M005 380-008-M025
89149-792EA 71.1 CAD
89149-792 89149-794 89149-796
Tacrolimus ≥98%, crystalline powder
Tacrolimus

Immunosuppressant.


  • Formulation: White to off-white crystalline powder.


Immunosuppressant that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2. Shown to inhibit the activity of FK506 binding protein and to inhibit the Ca²⁺-dependent phosphatase and protein phosphatase 2B (PP2B; calcineurin). Activates NF-κB through phosphorylation and degradation of IκBα.


Soluble in DMSO, 100% ethanol, methanol, ethyl acetate or acetone; insoluble in water.

Formula: C₄₄H₆₉NO₁₂
Storage Temperature: Freezer
MDL Number: MFCD00869853
CAS Number: 104987-11-3
UN: 2811
ADR: 6.1,III
Merck Index: 14,09025
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edab690b293d44c5d61333d1602b6d2639941289&vid=3b995985ecb62ed0dd2a7b2599972af120c9580c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (Assay)
Source Isolated from Streptomyces tsukubaensi


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR