Ponasterone A ≥95% (by HPLC)

Supplier: Enzo Life Sciences

370-014-M001 370-014-M005
89149-776EA 121.66 CAD
89149-776 89149-778 89149-780
Ponasterone A ≥95% (by HPLC)
Ponasterone A

Ecdysteroid.


  • Formulation: White to off-white powder.


A member of the ecdysteroid family. Analog of ecdysone with similar properties to muristerone A. It is a functional, reliable and economical substitute for muristerone A as an inducer for the ecdysone-inducible mammalian expression system. Results show that ponasterone A was able to induce expression of β-galactosidase to levels similar to those obtained with muristerone A induction. Potent compound for ecdysteroid activity; EC₅₀ = 0.07 µM.


Soluble in acetic acid, 100% ethanol, methanol or DMSO; insoluble in water.


Synonyms: 25-Deoxy-20-hydroxyecdysone; 25-deoxyecdysterone; 2β,3β,14α,20R,22R-pentahydroxy-5β-cholest-7-en-6-one

Formula: C₂₇H₄₄O₆
Storage Temperature: Freezer
CAS Number: 13408-56-5
Merck Index: 14,07588
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d541c4eda58e8b4c12036be83ab0842f13c858f1&vid=19a0e2fab2c15f1fd578bde5233c39811999655c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (HPLC)
Identity Identity determined by ¹H- and ¹³C-NMR
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR