20-Hydroxyecdysone ≥95% (by 1H-NMR), white powder

Supplier: Enzo Life Sciences

370-012-M005 370-012-M010 370-012-M050
89149-766EA 132.72 CAD
89149-766 89149-768 89149-770
20-Hydroxyecdysone ≥95% (by 1H-NMR), white powder
20-Hydroxyecdysone

Ecdysteroid.


  • Formulation: White powder.


Most widely occurring ecdysteroid in both plant and animal species. Controls cell death during metamorphosis of Drosophila melanogaster.


Soluble in water, 100% ethanol, methanol, DMSO and acetic acid.

Formula: C₂₇H₄₄O₇
Storage Temperature: Freezer
MDL Number: MFCD00036740
CAS Number: 5289-74-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da44659497a9327aef69595cf1596db1bafaadfb&vid=646b458542ef0550546fb47270640760eda15f35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (¹H-NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR