Genipin (from Gardenia jasminoides ellis) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

350-383-M025 350-383-M100
89149-650EA 146.94 CAD
89149-650 89149-652
Genipin (from Gardenia jasminoides ellis) ≥98% (by HPLC)
Genipin

Cell permeable inhibitor of uncoupling protein 2 (UCP2). Increases glucose-stimulated insulin secretion, mitochondrial membrane potential and ATP levels in pancreatic island cells. Induces apoptosis. Protein cross-linking agent. Anti-inflammatory and anti-angiogenic.


  • UCP2 inhibitor


White to off-white crystalline solid.


Soluble in DMSO (50mg/ml), dimethyl formamide (25mg/ml), 100% ethanol (5mg/ml), methanol or ethyl acetate; insoluble in water.

Formula: C₁₁H₁₄O₅
Storage Temperature: Freezer
MDL Number: MFCD00888600
CAS Number: 6902-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fe644bb56c9929d511d13f56509ee22a54b3f48&vid=4f5b3d9058d28f1774f5542382e974ccd0d4fd96&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR