3,5-Di-O-caffeoylquinic acid ≥97%

Supplier: Enzo Life Sciences

Synonyms: Isochlorogenic acid, (1R,3R,4S,5R)-3,5-Bis((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4-dihydroxy-3-methylcyclohexane-1-carboxylic acid, 3,5-CQA, Isochlorogenic acid A

350-320-M001 350-320-M005
89149-486EA 248.06 CAD
89149-486 89149-488
3,5-Di-O-caffeoylquinic acid ≥97%
3,5-Di-O-caffeoylquinic acid

Antioxidant.


  • Formulation: Off-white to yellow/brown crystalline solid.


Shows antiproliferative activity.


Soluble in DMSO or 100% ethanol.


Synonym: 3,5-CQA, Isochlorogenic acid

Formula: C₂₅H₂₄O₁₂
Storage Temperature: Freezer
MDL Number: MFCD00951290
CAS Number: 2450-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9bd9b89af4f178e863cc84b5af148aae298a8c4&vid=4d38ed7f5512a052f9324327bdf90c5a476e7efb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% by HPLC
Identity Determined by ¹H-NMR, ¹³C-NMR and MS
Source Isolated from Cynara scolymus


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR