Betulinic acid ≥99%, high purity
Supplier: Enzo Life Sciences
Synonyms:
(1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-Hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid, 3beta-Hydroxy-20(29)-lupaene-28-oic Acid
Antitumor and anti-HIV agent
Antitumor and anti-HIV agent. Induces apoptosis by activating mitochondrial permeability transition (MPT). Inhibits NF-κB activation and NF-κB-regulated gene expression induced by carcinogens and inflammatory stimuli. Decreases expression of Bcl-2 and cyclin D1. Potent proteasome activator. Triggers CD95- and p53-independent apoptosis via activation of caspases. It induces mitochondrial permeability pore opening and the release of cytochrome c and apoptosis-inducing factor. Displays anti-HIV activity. EC50 for inhibion of HIV-1 replicaton in H9 lymphocytes: 1.4µM.
Isolated from Platanus acerifolia (plane) tree bark.
Formula:
C₃₀H₄₈O₃ |
MDL Number:
MFCD00009619 CAS Number: 472-15-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...