(S)-(+)-Abscisic acid cis- and trans-mixture (from Curvularia lunata) ≥97% (by HPLC)
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
ABA, (+)-cis,trans ABA, (S)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-(2Z,4E)-pentadienoic acid, (S)-(+)-Abscisic acid, Dormin, S-ABA, (+)-Abscisic acid
350-255-C500
350-255-M001
89149-342EA
224.36
CAD
89149-342
89149-344
(S)-(+)-Abscisic acid cis- and trans-mixture (from Curvularia lunata) ≥97% (by HPLC)
(S)-(+)-Abscisic acid
Plant hormone.
- Formulation: Off-white powder.
Natural and active isomer of the abscission accelerating plant hormone.
Soluble in ethanol or DMSO.
Synonym: (+)-cis,trans ABA
Formula:
C₁₅H₂₀O₄ MW: 264.32 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00066545 CAS Number: 21293-29-8 Merck Index: 14,00011 |
Specification Test Results
Purity | ≥97% by HPLC |
Source | Isolated from Curvularia lunata |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...