Gramicidin A (from Bacillus brevis) ≥90% (by HPLC)

Supplier: Enzo Life Sciences

350-233-M001 350-233-M005
89149-300EA 88.48 CAD
89149-300 89149-302
Gramicidin A (from Bacillus brevis) ≥90% (by HPLC)
Gramicidin A

Ionophore antibiotic.


  • Contaminants: ≤5% Gramicidin C (HPLC)


Naturally occurring ion channel forming pentadecapeptide. Causes K+/H+-exchange in mitochondria in a non-voltage dependent manner.


White to off-white powder


Soluble in DMSO, lower alcohols or acetic acid


Packaging: Shipping temperature: Ambient

Formula: C₉₉H₁₄₀N₂₀O₁₇
Storage Temperature: Freezer
MDL Number: MFCD00466944
CAS Number: 11029-61-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR