Artemisinin ≥98%

Supplier: Enzo Life Sciences

350-219-G001 350-219-M100
89149-274EA 508.76 CAD
89149-274 89149-276
Artemisinin ≥98%
Artemisinin

iNOS inhibitor


Powerful antimalarial agent. Inhibits angiogenesis by down-regulating HIF-1α and VEGF expression in mouse embryonic stem cells. Crosses the blood-brain barrier. Inhibitor of human iNOS (NOS II).


Isolated from the traditional Chinese anti-malarial herb Artemisia annua L.

Formula: C₁₅H₂₂O₅
MDL Number: MFCD00081057
CAS Number: 63968-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42cc48aa25d46be973b0cfae1dbd7eb37e7d213c&vid=74809fe55986ae6423720d7d34e971884312c520&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR