Topotecan hydrochloride ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: SKF-104864A hydrochloride, 10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride, NSC 609669 hydrochloride, Topotecan HCl, 9-[(dimethylamino)methyl]-10-hydroxy-(20S)-camptothecin hydrochloride, (S)-10-((Dimethylamino)methyl)-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride

350-133-M001 350-133-M005 350-133-M025
89149-138EA 75.84 CAD
89149-138 89149-140 89149-142
Topotecan hydrochloride ≥98% (by HPLC)
Topotecan hydrochloride

Potent antitumor agent. Inhibitor of DNA-topoisomerase. Analog of (S)-(+)-camptothecin. Used in chemotherapy of several different cancers.


  • Topoisomerase I inhibitor


Light yellow to greenish powder.


Soluble in water (1mg/ml) or methanol.

Formula: C₂₃H₂₃N₃O₅.HCl
MW: 457.91 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866235
CAS Number: 119413-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59ab9fda4d83e9a357bcd4d39554794db090d337&vid=5ecd9e56fde6ecae9d21ae76bf8a5c02cf406aa1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR