Okadaic acid potassium salt ≥98% (by HPLC), high purity

Supplier: Enzo Life Sciences

350-063-C050 350-063-C100 350-063-M001
89148-998EA 237 CAD
89148-998 89149-000 89149-002
Okadaic acid potassium salt ≥98% (by HPLC), high purity
Okadaic acid potassium salt

Inhibitor of PP1 and PP2A


Salt form of okadaic acid (Prod. No. ALX-350-003), with slightly greater stability than the free acid after it is put into stock solution (in organic solvents). Potent inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). Potent tumor promotor and neurotoxin.


Isolated from Prorocentrum concavum by preparative flash, medium pressure and high performance liquid chromatography. Salt form generated in aqueous potassium hydroxide-methanol solution.

Formula: C₄₄H₆₇O₁₃·K
MDL Number: MFCD00214356
CAS Number: 155751-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=745844f0d4d838899a2f141d773b730aef7859fc&vid=76f38557a655a6f474376a277140a961b64b401a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR