(S)-(+)-Camptothecin ≥98%

Supplier: Enzo Life Sciences

Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

350-015-G001 350-015-M050 350-015-M250
89148-936EA 853.2 CAD
89148-936 89148-938 89148-940
(S)-(+)-Camptothecin ≥98%
(S)-(+)-Camptothecin

Potent antitumor agent


Potent antitumor agent. Inhibitor of DNA-topoisomerase I. Activates p53 resulting in upregulated expression of TRAIL-R2 (DR5) and Bak to overcome TRAIL resistance in Bax-deficient human colon carcinoma cells. Inhinbits Tat-mediated transactivation of HIV-1. Induces apoptosis in osteosarcoma and hepatoma cells. Suppresses nitric oxide (NO) biosynthesis.


Isolated from Mappia foetida Miers (Nothapodytes foetida (Wt.) Sleumer).

Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
MDL Number: MFCD00081076
CAS Number: 7689-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79e312ef261069ac395a1b5d7f1ba1b1194859ed&vid=8a5d8d23f288147c25f9ce8ed55d0dd1116276bb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR