Okadaic acid sodium salt ≥98% (by HPLC), high purity

Supplier: Enzo Life Sciences

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium salt, OA sodium salt

350-011-C025 350-011-C100 350-011-M001
89148-918EA 140.62 CAD
89148-918 89148-920 89148-922
Okadaic acid sodium salt ≥98% (by HPLC), high purity
Okadaic acid sodium salt

Inhibitor of PP1 and PP2A


Salt form of okadaic acid (Prod. No. ALX-350-003), with slightly greater stability than the free acid after it is put into stock solution (in organic solvents). Potent inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). Potent tumor promotor and neurotoxin.


Isolated from Prorocentrum concavum by preparative flash, medium pressure and high performance liquid chromatography. Salt form generated in aqueous sodium hydroxide-methanol solution.

Formula: C₄₄H₆₇NaO₁₃
MW: 827.00 g/mol
MDL Number: MFCD00210208
CAS Number: 209266-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cf174a5bd12de868c81223f9a5c29873dc698e6&vid=c1cc9da2275c9856dedabdc88e5e4983f3d30f5c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR