Okadaic acid ammonium salt ≥98% (by HPLC), high purity

Supplier: Enzo Life Sciences

350-010-C100 350-010-M001
89148-912EA 141 CAD
89148-912 89148-914 89148-916
Okadaic acid ammonium salt ≥98% (by HPLC), high purity
Okadaic acid ammonium salt

Inhibitor of PP1 and PP2A


Salt form of okadaic acid (Prod. No. ALX-350-003), with slightly greater stability than the free acid after it is put into stock solution (in organic solvents). Potent inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). Potent tumor promotor and neurotoxin.


Isolated from Prorocentrum concavum by preparative flash, medium pressure and high performance liquid chromatography. Salt form generated in aqueous ammonium hydroxide-methanol solution.

Formula: C₄₄H₆₇O₁₃·NH₄
MDL Number: MFCD00153856
CAS Number: 155716-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c9d1cd13df2f9113e1ff8c138f8cf669e4c0b3e&vid=d8096e6b6aa1785e33005f3ff82498443f98928b&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR