Ganglioside GD1a disodium salt ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: GD1a . 2Na

302-007-M001 302-007-M005
89148-650EA 135.88 CAD
89148-650 89148-652
Ganglioside GD1a disodium salt ≥98% (by TLC)
Ganglioside GD1a disodium salt

Differentiation marker for cell growth. Protein kinase C (PKC) inhibitor. The major ganglioside of the nervous system.


  • PKC inhibitor


Lyophilized.


Soluble in chloroform/methanol/DI water, 2:1:0.1

Formula: C₈₄H₁₄₆N₄O₃₉·2Na
MW: 1882.1 g/mol
Storage Temperature: Freezer
CAS Number: 12707-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7e7cf0d20000d70e3231ebda6f03b7027d73cd9&vid=f05bd540cc695a11afdaacddb0c1e8ee6b281106&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR