trans-Ned-19

Supplier: Enzo Life Sciences

270-503-M001 270-503-MC05
89148-118EA 699.94 CAD
89148-118 89148-120
trans-Ned-19
trans-Ned-19

Cell permeable, selective and potent inhibitor of nicotinic acid adenine dinucleotide phosphate (NAADP) receptor in mammalian cells and sea urchin eggs. Blocks NAADP signaling. Inhibits Ca2+ release: trans-Ned-19 (IC50 = 6nM); cis-Ned-19 (IC50 = 800nM). [32P] NAADP binding: trans-Ned-19 (IC50 = 0.4nM); cis-Ned-19 (IC50 = 15µM).
Inhibits Ca2+ release from lysosomes. Noncompetitive antagonist. Fluorescent.


Synthetic

Formula: C₃₀H₃₁FN₄O₃
MDL Number: MFCD02057367
CAS Number: 1354235-96-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5211ee023bb22c70e8b483cf4dc7bd5d3a95791f&vid=8fe12dc0d9bee4c9707257dd008ecc2162ac7a61&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR