Finasteride ≥98% (by UV method)

Supplier: Enzo Life Sciences

270-491-M100 270-491-M500
89148-076EA 165.9 CAD
89148-076 89148-078
Finasteride ≥98% (by UV method)
Finasteride

5alpha-reductase inhibitor


Inhibitor of 5α-reductase, the enzyme which converts testosterone to the more potent androgen, 5α-dihydrotestosterone. Used as a treatment in benign prostatic hyperplasia and prostate cancer. Also used to treat male pattern hair loss.

Formula: C₂₃H₃₆N₂O₂
MDL Number: MFCD00869737
CAS Number: 98319-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d2d7d0d5182c3f87ff1e6cdffd26f258c137355&vid=90b8d535f37578296b7c6db28e286f9777af076f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR