SB 225002 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

270-478-M001 270-478-M005
89148-040EA 75.84 CAD
89148-040 89148-042
SB 225002 ≥98% (by HPLC)
SB 225002

Chemokine receptor ligand


Potent and selective antagonist of CXCR2 chemokine receptor (IC50=22nM). >150-fold more selective over CXCR1 receptor. Inhibits IL-8- (IC50=8nM) and GROα-mediated (IC50=10nM) calcium mobilisation. Prevents IL-8-induced neutrophil migration. Inhibits HIV replication in lymphocytes and macrophages.

Formula: C₁₃H₁₀BrN₃O₄
MDL Number: MFCD00954637
CAS Number: 182498-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc0ca2f3e56a9482cc5dac2d8e903568ce9c6111&vid=6bad18652cde0412c1706e860b15d49941f5c8ea&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR