Bicalutamide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulphonyl)-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulphonyl]-2-hydroxy-2-methylpropionamide, N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulfonyl)-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropionamide

270-476-G001 270-476-M100 270-476-M500
89148-032EA 551.42 CAD
89148-032 89148-034 89148-036
Bicalutamide ≥98% (by HPLC)
Bicalutamide

Androgen receptor inhibitor


Nonsteroidal antiandrogen. Effective in the treatment of prostate cancer. Induces cell death by a pathway that is independent of changes in mitochondrial membrane potential and Bcl-2 action.


Synthetic.

Formula: C₁₈H₁₄F₄N₂O₄S
MW: 430.37 g/mol
MDL Number: MFCD00869971
CAS Number: 90357-06-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbbd0f5d7213c30414a27d6f8c25152f50b6ffdd&vid=4cb50b333010d4522505e142091cba339fb71e57&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR