PD 184,352 ≥95% (by HPLC, NMR)

Supplier: Enzo Life Sciences

270-471-M001 270-471-M005
89148-018EA 213.3 CAD
89148-018 89148-020
PD 184,352 ≥95% (by HPLC, NMR)
PD 184,352

Potent and selective inhibitor of MAPK (ERK kinase 1; MEK1) activation (IC50=300nM in vitro, IC50=2nM in vivo). Suppresses activation of MAPK but does not block its activity. Strongly suppresses growth of human implanted colon tumors in mice.


Synthetic.

Formula: C₁₇H₁₄ClF₂IN₂O₂
MDL Number: MFCD02683961
CAS Number: 212631-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e3b6b5e1369bd8fc76cd1d50bc412694525fac0&vid=688b38165cc8cd8faa5536352aa4bdad633d3be8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR