Sorafenib (BAY 43-9006) ≥95% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, Sorafenib, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)phenoxy)-N-methylpicolinamide

270-469-M001 270-469-M005 270-469-M025
89148-008EA 143.78 CAD
89148-008 89148-010 89148-012
Sorafenib (BAY 43-9006) ≥95% (by HPLC)
Sorafenib (BAY 43-9006)

Broad-spectrum kinase inhibitor


Broad-spectrum kinase inhibitor.


Synthetic.

Formula: C₂₁H₁₆ClF₃N₄O₃
MW: 464.82 g/mol
MDL Number: MFCD06411450
CAS Number: 284461-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b86126ae2f67d16a34b19c2fdfaf5712aa9708a1&vid=9e71960aa284c6661ef409ce0abedf8fd93b8840&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR