N-ω-Hydroxy-L-norarginine acetate salt ≥97% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: nor-NOHA acetate

270-467-M001 270-467-M005 270-467-M025
89148-000EA 83.74 CAD
89148-000 89148-002 89148-004
N-ω-Hydroxy-L-norarginine acetate salt ≥97% (by TLC)
N-ω-Hydroxy-L-norarginine acetate salt

Potent and reversible arginase inhibitor. Specifically interacts with the manganese-cluster of the arginase active site. Does not act as a substrate or as an inhibitor of NOS.


  • Arginase inhibitor


White to slightly yellowish powder.


Soluble in water (50mg/ml).

Formula: C₅H₁₂N₄O₃.CH₃COOH
MW: 236.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318777
CAS Number: 189302-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=086fd27b8eedf15dc606fc2137b02f72463d9c3e&vid=2b35cd137ba0c0214373d80a295913bed1e0e740&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR