Fluvastatin sodium salt ≥98% (by HPLC)

Supplier: Enzo Life Sciences

270-466-M010 270-466-M050
89147-996EA 115.34 CAD
89147-996 89147-998
Fluvastatin sodium salt ≥98% (by HPLC)
Fluvastatin sodium salt

Anticholesterol agent


Anticholesterol agent. HMG-CoA reductase inhibitor and antioxidant.


Synthetic.


HMG-CoA inhibitor.

Formula: C₂₄H₂₅FNNaO₄
MDL Number: MFCD00929076
CAS Number: 93957-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f01fd3cbc6e1c628def37442c08f4848d07f4cf&vid=501a5846f53bd1f8f56ec9658b3081894893b83a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR