AS-252424 ≥98% (by 1H-NMR)

Supplier: Enzo Life Sciences

270-465-M001 270-465-M005
89147-990EA 79 CAD
89147-990 89147-992 89147-994
AS-252424 ≥98% (by 1H-NMR)
AS-252424

Potent and selective phosphoinositide 3-kinase (PI(3)K) p110γ inhibitor shown to be selective for class IB PI(3)K-mediated cellular effects.

Formula: C₁₄H₈FNO₄S
MW: 305.3 g/mol
MDL Number: MFCD11045281
CAS Number: 900515-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7fb9fd4aa4bf505ea203ace315d31c05818368d&vid=c158e2b3881a96e5017b668f3817166b6bcaaee9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR