RO-3306 ≥95% (by 1H-NMR)

Supplier: Enzo Life Sciences

Synonyms: (Z)-5-(Quinolin-6-ylmethylene)-2-((thiophen-2-ylmethyl)amino)thiazol-4(5H)-one, (Z)-5-(Quinolin-6-ylmethylene)-2-[(thiophen-2-ylmethyl)amino]thiazol-4(5H)-one

270-463-M001 270-463-M005
89147-982EA 121.66 CAD
89147-982 89147-984
RO-3306 ≥95% (by 1H-NMR)
RO-3306

CDK/cyclin inhibitor


Inhibitor of CDK1/cyclin B1 and CDK1/cyclin A. Reversibly arrests human cells at the G2/M border of the cell cycle. Allows for effective cell synchronisation in early mitosis. Downregulates expression of the antiapoptotic proteins Bcl-2 and survivin and blocks p53-mediated induction of p21 and MDM2.

Formula: C₁₈H₁₃N₃OS₂
MW: 351.5 g/mol
MDL Number: MFCD17392573
CAS Number: 872573-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34b0bd01d1d143e4e067a94f3e7900d86378a898&vid=b30b0e437e91d615136d8ac17b5b0afe758525b8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR