RepSox ≥97% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-[3-(6-Methylpyridin-2-yl)-1H-pyrazol-4-yl]-1,5-naphthyridine, 2-(3-(6-Methylpyridin-2-yl)-1H-pyrazol-4-yl)-1,5-naphthyridine, ALK 5 Inhibitor II, TGF-beta RI Kinase Inhibitor II

270-445-M001 270-445-M005
89147-924EA 151.68 CAD
89147-924 89147-926
RepSox ≥97% (by HPLC)
RepSox

Cell permeable, potent, selective and ATP-competitive inhibitor of TGF-β RI kinase (IC50=23nM, 4nM and 18nM for binding, auto-phosphorylation and cellular assay in HepG2 cells of TGF-β RI kinase, respectively). Minimally affects a panel of 9 closely related kinases including p38 MAPK (IC50>16µM).


White to off-white or yellow solid.


5mg/ml soluble in DMSO or methanol.

Formula: C₁₇H₁₃N₅
MW: 287.326 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09037561
CAS Number: 446859-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca65bea25f291032efb27224677ce318435a4c41&vid=09f00f75ac9f61fb3122dbca40c32bae365f18d1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR