ABT-888 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (R)-2-(2-Methylpyrrolidin-2-yl)-1H-benzo[d]imidazole-7-carboxamide dihydrochloride, Veliparib 2HCl

270-444-M001 270-444-M005
89147-920EA 150.1 CAD
89147-920 89147-922
ABT-888 ≥98% (by HPLC)
ABT-888

PARP inhibitor


Potent inhibitor of PARP-1 and PARP-2 (potency ≤5nM in vitro). Does not inhibit other NAD-binding enzymes. Has minimal CYP450 inhibition and induction. Shows broad spectrum of chemo- and radiopotentiation. Is toxic to both oxic and hypoxic cells. Enantiomeric purity ≥97% suitable for in vivo studies. Does not show inherent cytotoxicity and shows no single agent activity in tumor models. Has excellent bioavailability and good blood-brain permeation. Increases tumor growth delay resulting from radiation and DNA-damaging agents.

Formula: C₁₃H₁₈Cl₂N₄O
MW: 317.214 g/mol
MDL Number: MFCD12407402
CAS Number: 912445-05-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0283cb8fe5c89236d4b3e2cc31a9a41ef442deb&vid=a5eeefbd3ffe71bf46f76be5ff54aac8399b2c79&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR